Anti-CYLC2 Rabbit Polyclonal Antibody

Supplier: Proteintech
18729-1-AP-150UL
10085-492EA 402 USD
10085-492
Anti-CYLC2 Rabbit Polyclonal Antibody
Antibodies
CYLC2, also named as CYL2, possible plays an architectural role during spermatogenesis. It may be involved in spermatid differentiation.

Western Blot: Human adrenal gland Tissue, 1:200-1:2000; IHC: Human Testis, 1:20-1:200

Type: Primary
Antigen: CYLC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR