Anti-GABARAP Rabbit Polyclonal Antibody

Supplier: Proteintech
18723-1-AP-150UL
10087-222EA 402 USD
10087-222
Anti-GABARAP Rabbit Polyclonal Antibody
Antibodies
GABARAP (Gamma-aminobutyric acid type A receptor-associated protein), was initially identified as GABAA receptor binding protein and has shown to be implicated in intracellular trafficking of GABAA receptors. Through interacting with a broad rang of proteins, GABARAP has been demonstrated as a multifunctional protein implicated in intracellular vesicle trafficking, apoptosis, and autophagy. The protein level of lipidated form of GABARAP (GABARAP-II) is upregulated during the autophagic process, which makes it as an autophagic marker. This antibody detected endogenous GABARAP protein in lysates from various tissues. It may cross-react with GABARAPL1 due to the high identity of amino acid sequence.

Western Blot: Human Placenta Tissue, 1:500-1:5000

Type: Primary
Antigen: GABARAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR