Anti-Creatine kinase B type-Specific Rabbit Polyclonal Antibody

Supplier: Proteintech
18713-1-AP-150UL
10082-026EA 402 USD
10082-026
Anti-Creatine kinase B type-Specific Rabbit Polyclonal Antibody
Antibodies
CKBB, also named as B-CK and CKB, is a member of the ATP:guanido phosphotransferase protein family. It is a cytoplasmic enzyme involved in energy homeostasis. CKBB reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. CK isoenzymes play a central role in energy transduction in tissues with large, fluctuating energy demands, such as skeletal muscle, heart, brain and spermatozoa. CK MB consists of a dimer of nonidentical chains. With MM being the major form in skeletal muscle and myocardium, MB existing in myocardium, and BB existing in many tissues, especially brain.

Western Blot: HEK-293 Cells, 1:1000-1:10000; IHC: Human gliomas, 1:20-1:200

Type: Primary
Antigen: Creatine kinase B type-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR