Anti-CPS1 Rabbit Polyclonal Antibody

Supplier: Proteintech
18703-1-AP-150UL
10082-050EA 402 USD
10082-050
Anti-CPS1 Rabbit Polyclonal Antibody
Antibodies
CPS1 is an enzyme that catalyzes the first committed step of the hepatic urea cycle, which is important in the removal of excess urea from cells. Expressed primarily in the liver and small intestine, CPS1 can be used as a marker for mitochondria in these tissues. In addition, CPS1 expression appears to be lost in adenocarcinomas of the small intestine. The antibody can recognize two isoforms of this gene around 165 kDa and 116 kDa.

Western Blot: Mouse Liver Tissue, 1:1000-1:10000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: CPS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR