Anti-NUMB Rabbit Polyclonal Antibody

Supplier: Proteintech
18701-1-AP-150UL
10081-684EA 402 USD
10081-684
Anti-NUMB Rabbit Polyclonal Antibody
Antibodies
NUMB, also named as S171, plays a role in the determination of cell fates during development. It is a membrane-bound protein that has been shown to associate with EPS15, LNX1, and NOTCH1. The degradation of NUMB is induced in a proteasome-dependent manner by MDM2. Four transcript variants encoding different isoforms have been found for this gene. The antibody can bind to all these four isoforms P65, P66, P71 and P72, can’t recognize the NUMBL.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human Brain Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; FC: HepG2 Cells, N/A

Type: Primary
Antigen: NUMB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR