Anti-SRA1 Rabbit Polyclonal Antibody

Supplier: Proteintech
18699-1-AP-150UL
10081-530EA 402 USD
10081-530
Anti-SRA1 Rabbit Polyclonal Antibody
Antibodies
Steroid receptor RNA activator 1 (SRA1), also known as seroid receptor RNA activator protein (SRAP) or SRA, is involved in modulating the activity of multiple transcription factors including the estrogen receptor (ER) . It may play a role in tumorigenesis . The SRA1 gene encodes both SRA1 protein and a non-coding functional RNA that functions as part of a ribonucleoprotein complex activating steroid receptor induced transcription.

Western Blot:A431 Cells, 1:200-1:2000

Type: Primary
Antigen: SRA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR