Anti-BRK Rabbit Polyclonal Antibody

Supplier: Proteintech
18697-1-AP-150UL
10083-686EA 402 USD
10083-686
Anti-BRK Rabbit Polyclonal Antibody
Antibodies
BRK, also named as PTK6 and FLJ42088, is a cytoplasmic nonreceptor protein kinase which may function as an intracellular signal transducer in epithelial tissues. PTK6 can phosphorylate KHDRBS1, KHDRBS2, KHDRBS3 and STAP2/BKS. Overexpression of PTK6 in mammary epithelial cells leads to sensitization of the cells to epidermal growth factor and results in a partially transformed phenotype. Expression of PTK6 has been detected at low levels in some breast tumors but not in normal breast tissue. Its presence in the nucleus appears to be linked to suppression of tumor progression. This antibody is specific to PTK6.

Western Blot: MCF7 Cells, 1:500-1:5000; IHC: Human Colon Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:10-1:100

Type: Primary
Antigen: BRK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR