Anti-KIT Rabbit Polyclonal Antibody

Supplier: Proteintech
18696-1-AP-150UL
10084-802EA 402 USD
10084-802
Anti-KIT Rabbit Polyclonal Antibody
Antibodies
KIT, also named as SCFR, c-Kit and CD117, is a transmembrane tyrosine kinase encoded by the cKit proto oncogene. It is a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor). KIT acts to regulate a variety of biological responses including cell proliferation, apoptosis, chemotaxis and adhesion. Ligand(SCF) binding to the extracellular domain leads to autophosphorylation on several tyrosine residues within the cytoplasmic domain, and activation. Phosphorylation at tyrosine 721 of KIT allows binding and activation of PI3 kinase. Loss of expression of KIT appears to be associated with progression of some tumors (melanoma) and autocrine/paracrine stimulation of the kit/SCF system may participate in human solid tumors such as lung, breast, testicular and gynecological malignancies. Mutations in Kit have been found to be important for tumor growth and progression in a variety of cancers including mast cell diseases, gastrointestinal stromal tumor, acute myeloid leukemia, Ewing sarcoma and lung cancer.

Western Blot: MCF7 Cells, 1:200-1:1000; IHC: Human Testis Tissue, 1:20-1:200; FC: MCF-7 Cells, N/A

Type: Primary
Antigen: KIT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR