Anti-TSPAN7 Rabbit Polyclonal Antibody

Supplier: Proteintech
18695-1-AP-150UL
10096-442EA 402 USD
10096-442
Anti-TSPAN7 Rabbit Polyclonal Antibody
Antibodies
TSPAN7, also named as A15, MXS1, CD231, MRX58, CCG-B7, TM4SF2, TALLA-1, TM4SF2b and DXS1692E, is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. TSPAN7 is a cell surface glycoprotein and may have a role in the control of neurite outgrowth. TSPAN7 may be involved in cell proliferation and cell motility. It is not solely expressed in T cells, but also expressed in acute myelocytic leukemia cells of some patients. The antibody only recognize the protein TSPAN7, can’t recognize the protein TSPAN6.

Western Blot:K-562 Cells, 1:200-1:2000; IHC: Human spleen Tissue, 1:20-1:200

Type: Primary
Antigen: TSPAN7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR