Anti-FUT10 Rabbit Polyclonal Antibody

Supplier: Proteintech
18660-1-AP-150UL
10087-188EA 402 USD
10087-188
Anti-FUT10 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Testis, 1:20-1:200

Type: Primary
Antigen: FUT10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR