Anti-CLCN1 Rabbit Polyclonal Antibody

Supplier: Proteintech
18595-1-AP-150UL
10084-838EA 402 USD
10084-838
Anti-CLCN1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CLCN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR