Anti-GAL3ST1 Rabbit Polyclonal Antibody

Supplier: Proteintech
18503-1-AP-150UL
10087-320EA 402 USD
10087-320
Anti-GAL3ST1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Testis Tissue, 1:500-1:5000

Type: Primary
Antigen: GAL3ST1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR