Anti-CACNG5 Rabbit Polyclonal Antibody

Supplier: Proteintech
18498-1-AP-150UL
10083-944EA 402 USD
10083-944
Anti-CACNG5 Rabbit Polyclonal Antibody
Antibodies
Western Blot:SH-SY5Y Cells, 1:200-1:2000

Type: Primary
Antigen: CACNG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR