Anti-IKBKG Rabbit Polyclonal Antibody

Supplier: Proteintech
18474-1-AP-150UL
10088-688EA 402 USD
10088-688
Anti-IKBKG Rabbit Polyclonal Antibody
Antibodies
IKBKG, also named as FIP3, NEMO, IKKAP1 and IKKG, is specifically phosphorylate serine or threonine residues that are followed by a proline residue. IKBKG is regulatory subunit of the IKK core complex which phosphorylates inhibitors of NF-kappa-B thus leading to the dissociation of the inhibitor/NF-kappa-B complex and ultimately the degradation of the inhibitor. Its binding to scaffolding polyubiquitin seems to play a role in IKK activation by multiple signaling receptor pathways. IKBKG is a predominant 48-kD protein and an N-terminally truncated protein of 45 kDa produced in smaller amounts and translated from methionine-38.

Western Blot:K-562 Cells, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: IKBKG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR