Anti-PIAS4 Rabbit Polyclonal Antibody

Supplier: Proteintech
18469-1-AP-150UL
10081-364EA 402 USD
10081-364
Anti-PIAS4 Rabbit Polyclonal Antibody
Antibodies
PIAS4(protein inhibitor of activated STAT protein 4) is also named as PIASG, PIASy and belongs to the PIAS family. It enhanced the conjugation of SUMO2 to GATA2 through its E3 SUMO ligase activity. TGF-b can up-regulates PLAS4 expression in MCs to down-regulatea-SMA gene transcription by the interaction with E12 and PLAS4 predominantly acts as E3 ligases for E12 in MCs among PIAS family members. This protein can be sumoylated and Lys-35 is the main site of sumoylation.

Western Blot: HL-60 Cells, 1:500-1:5000

Type: Primary
Antigen: PIAS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR