Anti-FAM84B Rabbit Polyclonal Antibody

Supplier: Proteintech
18421-1-AP-150UL
10086-840EA 524.48 USD
10086-840
Anti-FAM84B Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:200-1:1000; IHC: Human oesophagus cancer, 1:20-1:200

Type: Primary
Antigen: FAM84B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR