Anti-P62 Rabbit Polyclonal Antibody

Supplier: Proteintech
18420-1-AP-150UL
10081-696EA 537.7 USD
10081-696
Anti-P62 Rabbit Polyclonal Antibody
Antibodies
Sequestosome 1 (SQSTM1/p62) is a multifunctional adaptor protein implicated in selective autophagy, cell signaling pathways, and tumorigenesis. p62 has been implicated in shuttling ubiquitinated and sometimes aggregated proteins for autophagic degradation. As a autophagy-specific substrate, p62 is degraded during the autophagic process, which makes intracellular level of p62 as a marker for autophagy flux. p62 is at the cross-roads of several signaling pathways including Ras/ Raf/ MAPK and NFκB and plays important role in cancer. p62 is a component of inclusion bodies/ protein aggregates found in human diseases, including Huntington's disease, Alzheimer's disease, Parkinson's disease in the brain, and nephropathic cystinosis in kidney (22074114, 22860231, 22714671). The molecular weight of p62 is predicted as 48/ 38 kDa, while western blot analyses using this antibody demonstrate the major band around 60-62 kDa in various tissues.

Western Blot: HEK-293 Cells, 1:1000-1:10000; IHC: Human gliomas Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200; IP: HEK-293 Cells, 1:1000-1:10000; FC: HEK-293T Cells, N/A

Type: Primary
Antigen: P62
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR