Anti-KISS1 Rabbit Polyclonal Antibody

Supplier: Proteintech
18375-1-AP-150UL
10089-288EA 537.7 USD
10089-288
Anti-KISS1 Rabbit Polyclonal Antibody
Antibodies
KISS1, also known as kisspeptin, is a metastasis suppressor protein that suppresses metastases of human melanomas and breast carcinomas without affecting tumorigenicity. It may regulate events downstream of cell-matrix adhesion, perhaps involving cytoskeletal reorganization. A C-terminally amidated peptide of KISS1, metastin (kisspeptin-54 or kp-54), functions as the endogenous ligand of the G-protein coupled receptor GPR54 . KISS1-GPR54 signaling is involved in the regulation of reproduction, playing an important role in initiating secretion of gonadotropin-releasing hormone (GnRH) at puberty .

Western Blot:NIH/3T3 Cells, 1:200-1:1000; IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: KISS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR