Anti-HNRNPL Rabbit Polyclonal Antibody

Supplier: Proteintech
18354-1-AP-150UL
10088-388EA 402 USD
10088-388
Anti-HNRNPL Rabbit Polyclonal Antibody
Antibodies
Heterogeneous nuclear ribonucleoprotein L (hnRNP L) is a component of the heterogeneous nuclear ribonucleoprotein (hnRNP) complexes which provide the substrate for the processing events that pre-mRNAs undergo before becoming functional, translatable mRNAs in the cytoplasm. It is associated with most nascent transcripts including those of the landmark giant loops of amphibian lampbrush chromosomes. The two isoforms of hnRNP L are generated by alternative splicing.The human hnRNPL gene encodes a protein that is localized in the nucleoplasm and cytoplasm. This antibody may recognize both isoforms of the hnRNP L protein.

Western Blot: RAW 264.7 Cells, 1:200-1:2000; IF: HeLa Cells, 1:10-1:100; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: HNRNPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR