Anti-KLF13 Rabbit Polyclonal Antibody

Supplier: Proteintech
18352-1-AP-150UL
10089-302EA 402 USD
10089-302
Anti-KLF13 Rabbit Polyclonal Antibody
Antibodies
Kruppel-like factor 13(KLF13) is a member of transcription actors, which contains 3 zinc finger DNA-binding domains. Theses transcription factors bind to CG-rich sequences and related GT and CACCC boxes. KLF13 involved in activation of transcription of variety of genes in the cells of erythroid lineage, such as GATA1, glycophorin B. It also binds to the A and A/B RANTES promoter binding sites, the same as NFKB, and then transactivates the RANTES gene.

Western Blot:L02 Cells, 1:200-1:2000; IHC: Human Brain, 1:20-1:200

Type: Primary
Antigen: KLF13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR