Anti-DTX1 Rabbit Polyclonal Antibody

Supplier: Proteintech
18350-1-AP-150UL
10086-156EA 402 USD
10086-156
Anti-DTX1 Rabbit Polyclonal Antibody
Antibodies
Deltex homolog 1 (E3 ubiquitin-protein ligase DTX1) is a regulator of Notch signaling, a signaling pathway involved in cell-cell communications that regulates a broad spectrum of cell-fate determinations. Probably acts both as a positive and negative regulator of Notch, depending on the developmental and cell context. Functions as an ubiquitin ligase protein in vivo, mediating ubiquitination and promoting degradation of MEKK1, suggesting that it may regulate the Notch pathway via some ubiquitin ligase activity.DTX1 is conserved from Drosophila, and is expressed in a wide array of tissues, particularly in the central nervous system, testes, and endothelial cells, mainly colocalized with Notch1.It contains a RING-type zinc finger and 2 WWE domains. This antibody recognizes DTX1 protein (72kd) and it's isoforms (about 60kd).

IF: Mouse olfactory epithelium Tissue, N/A; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: DTX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR