Anti-p35 Rabbit Polyclonal Antibody

Supplier: Proteintech
18346-1-AP-150UL
10091-680EA 402 USD
10091-680
Anti-p35 Rabbit Polyclonal Antibody
Antibodies
Cyclin dependent kinase-5 (Cdk5), is an atypical but essential member of the Cdk family of proline-directed serine/threonine kinases. CDK5R1 encodes p35, a specific activator of the serine/threonine kinase CDK5, which plays a central role in neuronal migration and differentiation during central nervous system development. The p35 form of this protein is proteolytically cleaved by calpain, generating a p25 form.

Western Blot: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: p35
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR