Anti-ITM2A Rabbit Polyclonal Antibody

Supplier: Proteintech
18306-1-AP-150UL
10081-588EA 537.7 USD
10081-588
Anti-ITM2A Rabbit Polyclonal Antibody
Antibodies
ITM2A, also named BRICD2A and E25A, is a member of type II integral membrane (ITM2) protein family. ITM2A expression is high in osteogenic and lymphoid tissues, while both ITM2B and ITM2C are expressed in brain. There are 5 articles specifically referring to this gene in PubMed. Proteins are expected to localize in various compartments (cytoplasm, integral to membrane). No phenotype has yet been reported to our knowledge: this gene's in vivo function is yet unknown.

IHC: Human Liver, 1:20-1:200

Type: Primary
Antigen: ITM2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR