Anti-ANG Rabbit Polyclonal Antibody

Supplier: Proteintech
18302-1-AP-150UL
10082-878EA 402 USD
10082-878
Anti-ANG Rabbit Polyclonal Antibody
Antibodies
Angiogenin (ANG), a angiogenic ribonuclease, is a member of the vertebrate-specific, secreted RNASE superfamily. Angiogenin, originally identified as a tumor angiogenic factor, was related with the growth and metastasis of numerous tumors. Angiogenin is also a component of normal human plasma and follicular Xuid. Its widespread expression in human tissues suggests angiogenin may have biological functions in addition to the promotion of angiogenesis. Recently, its biological activity has been extended to stimulating cell proliferation and promoting cell survival. It is a 17 kDa precursor which is cleaved to generate the 14 kDa mature protein.

Western Blot: Human vein Tissue, 1:200-1:2000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: ANG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR