Anti-Endostatin Rabbit Polyclonal Antibody

Supplier: Proteintech
18301-1-AP-150UL
10086-520EA 402 USD
10086-520
Anti-Endostatin Rabbit Polyclonal Antibody
Antibodies
Endostatin is a naturally-occurring 20-kDa C-terminal fragment derived from COL18A1 (also named as KNO). It is reported to serve as an anti-angiogenic agent, similar to angiostatin and thrombospondin. It potently inhibits endothelial cell proliferation and inhibits endothelial cell angiogenesis by binding to the heparan sulfate proteoglycans involved in growth factor signaling. Endostatin is a broad spectrum angiogenesis inhibitor and may interfere with the pro-angiogenic action of growth factors such as basic fibroblast growth factor (bFGF/FGF-2) and vascular endothelial growth factor (VEGF).

Western Blot: Human Liver Tissue, 1:200-1:1000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: Endostatin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR