Anti-Plasminogen Rabbit Polyclonal Antibody

Supplier: Proteintech
18300-1-AP-150UL
10081-382EA 402 USD
10081-382
Anti-Plasminogen Rabbit Polyclonal Antibody
Antibodies
Angiostatin is a part gene (79-466aa) of Plasminogen (PLG, 810aa). Angiostatin is an angiogenesis inhibitor that blocks neovascularization and growth of experimental primary and metastatic tumors in vivo. Angiostatin has 4 kringle domains.

Western Blot: Human serum Tissue, 1:200-1:1000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: Plasminogen
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR