Anti-BMF Rabbit Polyclonal Antibody

Supplier: Proteintech
18298-1-AP-150UL
10083-284EA 402 USD
10083-284
Anti-BMF Rabbit Polyclonal Antibody
Antibodies
BMF belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities such as central regulators of apoptotic cell death. BMF contains a single BCL2 homology domain 3 (BH3), and has been shown to bind BCL2 proteins and function as an apoptotic activator. This protein is found to be sequestered to myosin V motors by its association with dynein light chain 2, which may be important for sensing intracellular damage and triggering apoptosis. Alternatively spliced transcript variants encoding different isoforms have been identified. BMF is widely expressed in tissues such as pancreas, liver and kidney, and in hematopoietic tissues.

Western Blot: Human Placenta Tissue, 1:200-1:2000

Type: Primary
Antigen: BMF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR