Anti-HRAS Rabbit Polyclonal Antibody

Supplier: Proteintech
18295-1-AP-150UL
10088-448EA 537.7 USD
10088-448
Anti-HRAS Rabbit Polyclonal Antibody
Antibodies
HRAS,also named as c-bas/has and RASH1, is v-Ha-ras Harvey rat sarcoma viral oncogene homolog from the mammalian ras gene family and it is a member of the small GTPase superfamily. RAS proteins are involved in signal transduction pathways, and bind GDP/GTP and possess intrinsic GTPase activity. Defects in HRAS are a cause of cancer. The antibody is special to HRAS, can’t recognize NRAS and KRAS.

Western Blot: HEK-293 Cells, 1:200-1:1000; IHC: Human Kidney Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; FC: HeLa Cells, N/A

Type: Primary
Antigen: HRAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR