Anti-KCNE4 Rabbit Polyclonal Antibody

Supplier: Proteintech
18289-1-AP-150UL
10089-128EA 402 USD
10089-128
Anti-KCNE4 Rabbit Polyclonal Antibody
Antibodies
KCNE4, also named as MIRP3 and MGC2035, is a membrane protein belongs to a family of single transmembrane domain proteins known to have dramatic effect on the gating of certain potassium channels. KCNE4 is expressed strongly in heart, skeletal muscle and kidney. Electrophysiological studies show that human KCNE4 modulates the activation of the KCNQ1 channel.

Western Blot: Human Skeletal Muscle Tissue, 1:200-1:2000

Type: Primary
Antigen: KCNE4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR