Anti-TDP43 Rabbit Polyclonal Antibody

Supplier: Proteintech
18280-1-AP-150UL
10081-154EA 402 USD
10081-154
Anti-TDP43 Rabbit Polyclonal Antibody
Antibodies
Transactivation response (TAR) DNA-binding protein of 43 kDa (also known as TARDBP or TDP-43) was first isolated as a transcriptional inactivator binding to the TAR DNA element of the HIV-1 virus. Neumann et al. (2006) found that a hyperphosphorylated, ubiquitinated, and cleaved form of TARDBP, known as pathologic TDP-43, is the major component of the tau-negative and ubiquitin-positive inclusions that characterize amyotrophic lateral sclerosis (ALS) and the most common pathological subtype of frontotemporal lobar degeneration (FTLD-U). 18280-1-AP is a rabbit polyclonal antibody raised against the full-length of human TDP-43. This antibody recognizes the cleavage product of 20-30 kDa in addition to the native and phosphorylated forms of TDP-43. Immunohistochemical analyses of TDP-43 using this antibody detect both normal diffuse nuclear staining and insoluble inclusions in pathologic tissues.

Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human gliomas Tissue, 1:20-1:200; IP:K-562 Cells, 1:500-1:5000

Type: Primary
Antigen: TDP43
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR