Anti-ATP6V0D1 Rabbit Polyclonal Antibody

Supplier: Proteintech
18274-1-AP-150UL
10083-360EA 402 USD
10083-360
Anti-ATP6V0D1 Rabbit Polyclonal Antibody
Antibodies
ATP6V0D1(V-type proton ATPase subunit d 1) is also named as ATP6D, VPATPD and belongs to the V-ATPase V0D/AC39 subunit family.It is responsible for acidifying a variety of intracellular compartments in eukaryotic cells, thus providing most of the energy required for transport processes in the vacuolar system.

Western Blot: Mouse Testis Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200; FC: HeLa Cells, N/A; IF: HeLa Cells, 1:20-1:200

Type: Primary
Antigen: ATP6V0D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR