Anti-PDPK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
18262-1-AP-150UL
10091-830EA 402 USD
10091-830
Anti-PDPK1 Rabbit Polyclonal Antibody
Antibodies
PDK1(pyruvate dehydrogenase kinase isoform 1) is also named as PDHK1 and belongs to the PDK/BCKDK protein kinase family.It inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.Starvation increases islet PDK activity.This antibody is speicfic to PDK1.

Western Blot: Mouse Liver Tissue, 1:200-1:2000; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: PDPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR