Anti-CHN2 Rabbit Polyclonal Antibody

Supplier: Proteintech
18259-1-AP-150UL
10084-728EA 402 USD
10084-728
Anti-CHN2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse cerebellum Tissue, 1:200-1:2000

Type: Primary
Antigen: CHN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR