Anti-EIF2AK2 Rabbit Polyclonal Antibody

Supplier: Proteintech
18244-1-AP-150UL
10086-346EA 537.7 USD
10086-346
Anti-EIF2AK2 Rabbit Polyclonal Antibody
Antibodies
The eukaryotic translation initiation factor 2-alpha kinase 2 (EIF2AK2, also known as PKR or PRKR) is an important component of the host innate immuneantiviral response.The product of the HSAEIF2AK2 gene is a 68 kDa protein that is ubiquitously expressed at low levels. The N-terminal part of this protein contains two dsRNA binding motifs (dsRBM) .The C-terminal EIF2S1 kinase domain includes 12 sub-domains that are conserved in several Species.This antibody is specific to EIF2AK2.

Western Blot: MCF7 Cells, 1:1000-1:10000; IHC: Human Testis, 1:20-1:200; IF: MCF-7, 1:10-1:100

Type: Primary
Antigen: EIF2AK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR