Anti-CHST3 Rabbit Polyclonal Antibody

Supplier: Proteintech
18242-1-AP-150UL
10084-770EA 402 USD
10084-770
Anti-CHST3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Heart Tissue, 1:200-1:2000; IHC: Human breast cancer, 1:20-1:200

Type: Primary
Antigen: CHST3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR