Anti-MMP28 Rabbit Polyclonal Antibody

Supplier: Proteintech
18237-1-AP-150UL
10090-304EA 402 USD
10090-304
Anti-MMP28 Rabbit Polyclonal Antibody
Antibodies
MMP28(Epilysin)can degrade casein and play a role in tissues homeostasis and repair .The only specific human cell type recognized to express epilysin is the basal keratinocyte in the skin, where epilysin expression is upregulated during wound repair.Western blot showed this antibody recognize bands at 62 and 58 kDa as well as breakdown products at 50, 48, and 46 kDa.This antibody is specific to MMP28.

Western Blot:A549 Cells, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: MMP28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR