Anti-CYP3A4 Rabbit Polyclonal Antibody

Supplier: Proteintech
18227-1-AP-150UL
10085-532EA 402 USD
10085-532
Anti-CYP3A4 Rabbit Polyclonal Antibody
Antibodies
Cytochrome P450 3A(CYP3A) belongs to the cytochrome P450 family and the enzymes constitute an important detoxification system that contributes to primary metabolism of more than half of all prescribed medications.CYP3A expression determines impairment of drug absorption and efficient systemic clearance in a tissue-specific manner.

Western Blot: Mouse Kidney Tissue, 1:200-1:2000; IHC: Human Liver Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: CYP3A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR