Anti-CCL28 Rabbit Polyclonal Antibody

Supplier: Proteintech
18214-1-AP-150UL
10084-216EA 402 USD
10084-216
Anti-CCL28 Rabbit Polyclonal Antibody
Antibodies
CCL28, also named as SCYA28, CCK1 and MEC, belongs to the intercrine beta (chemokine CC) family. It has chemotactic activity for resting CD4, CD8 T-cells and eosinophils. CCL28 binds to CCR3 and CCR10 and induces calcium mobilization in a dose-dependent manner. CCL28 is expressed in a variety of human and mouse tissues, and it appears to be predominantly produced by epithelial cells. It is produced by epithelial cells of these tissues suggesting that this chemokine can play an important role by linking homing mechanisms between the gut, nasal mucosa and mammary gland (MG). In WB test, CCL28 is 14kd and 8-9kd(a putatively to a degradation fragment).

Western Blot: Human spleen Tissue, 1:200-1:2000; IHC: Human Colon Tissue, 1:20-1:200

Type: Primary
Antigen: CCL28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR