Anti-PPM1L Rabbit Polyclonal Antibody

Supplier: Proteintech
18203-1-AP-150UL
10092-672EA 402 USD
10092-672
Anti-PPM1L Rabbit Polyclonal Antibody
Antibodies
PPM1L(Protein phosphatase 1L) is also named as PP2CE.It belongs to the PP2C group of serine/threonine phosphatases, which are distinguished from other phosphatases by their structure, absolute requirement for Mg(2+) or Mn(2+), and insensitivity to okadaic acid. It has 4 isoforms produced by alternative splicing.This antibody is specific to PPM1L.

Western Blot: Mouse Kidney Tissue, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: PPM1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR