Anti-LACTB Rabbit Polyclonal Antibody

Supplier: Proteintech
18195-1-AP-150UL
10089-468EA 402 USD
10089-468
Anti-LACTB Rabbit Polyclonal Antibody
Antibodies
LACTB is also named as MRPL56,whic is a component of the large subunit of the mitochondrial ribosome that is encoded by the nuclear genome.LACTB is a mammalian active-site serine protein that has evolved from a bacterial penicillin-binding protein.It promotes intramitochondrial membrane organization and micro-compartmentalization through polymerization. Northern blot analysis of mouse tissues detected expression at high levels in liver, heart, and skeletal muscle, with lower levels in kidney, testis, brain, lung, and spleen.This antibody is specific to LACTB.

Western Blot: Human Heart Tissue, 1:1000-1:10000; IHC: Human Liver, 1:20-1:200

Type: Primary
Antigen: LACTB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR