Anti-CMA Rabbit Polyclonal Antibody

Supplier: Proteintech
18189-1-AP-150UL
10084-896EA 402 USD
10084-896
Anti-CMA Rabbit Polyclonal Antibody
Antibodies
Mast Cell Chymase, also named as CYH, CYM and CMA1, belongs to the peptidase S1 family and Granzyme subfamily. It is the major secreted protease of mast cells with suspected roles in vasoactive peptide generation, extracellular matrix degradation, and regulation of gland secretion. CMA1 maybe a target for cardiovascular disease therapies.

Western Blot: Mouse Heart Tissue, 1:200-1:2000

Type: Primary
Antigen: CMA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR