Anti-ABCD2 Rabbit Polyclonal Antibody

Supplier: Proteintech
18179-1-AP-150UL
10082-346EA 402 USD
10082-346
Anti-ABCD2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse cerebellum Tissue, 1:200-1:1000; IP:A431 Cells, 1:200-1:1000

Type: Primary
Antigen: ABCD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR