Anti-CLEC4G Rabbit Polyclonal Antibody

Supplier: Proteintech
18173-1-AP-150UL
10084-858EA 402 USD
10084-858
Anti-CLEC4G Rabbit Polyclonal Antibody
Antibodies
Western Blot: HUVEC Cells, 1:200-1:1000

Type: Primary
Antigen: CLEC4G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR