Anti-FGF5 Rabbit Polyclonal Antibody

Supplier: Proteintech
18171-1-AP-150UL
10086-974EA 524.48 USD
10086-974
Anti-FGF5 Rabbit Polyclonal Antibody
Antibodies
Western Blot:NIH/3T3 Cells, 1:200-1:2000; IHC: Human Stomach Tissue, 1:20-1:200

Type: Primary
Antigen: FGF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR