Anti-AMPK alpha 2 Rabbit Polyclonal Antibody

Supplier: Proteintech
18167-1-AP-150UL
10082-854EA 402 USD
10082-854
Anti-AMPK alpha 2 Rabbit Polyclonal Antibody
Antibodies
PRKAA2(protein kinase, AMP-activated, alpha 2 catalytic subunit), also named as AMPKA2, AMPK, PRKAA, AMPK2, belonogs to the CAMK Ser/Thr protein kinase family and SNF1 subfamily. PRKAA2 is an αβγ heterotrimer that is activated by low cellular energy status, such as decreases in both the ATP/AMP ratio and the phosphocreatine content and it is a glycogen synthase kinase, phosphorylating Ser7 at the NH2 terminus, which decreases glycogen synthase activity. The protein can be ubiquitinated.

Western Blot: Human Skeletal Muscle Tissue, 1:1000-1:10000; IF: MCF-7 Cells, 1:10-1:100; IHC: Human Skeletal Muscle Tissue, 1:20-1:200

Type: Primary
Antigen: AMPK alpha 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR