Anti-MMP13 Rabbit Polyclonal Antibody

Supplier: Proteintech
18165-1-AP-150UL
10089-672EA 402 USD
10089-672
Anti-MMP13 Rabbit Polyclonal Antibody
Antibodies
MMP-13(Matrix metalloproteinase-13), also known as collagenase-3, cleaves type I collagen and was previously thought to be the rodent analogue of human interstitial collagenase (MMP-1). MMP-13 is found in hypertrophic and calcifying cartilage of mammalian growth plate. The bands seen with rat MMP-13 are consistent with a 59-kDa latent form and a 43-kDa active form reported for this enzyme ,along with a smaller band at approximately 30 kDa. It also can be detected 3 bands that the glycosylated proMMP-13 (70 kDa), unglycosylated proform (55 kDa), and active MMP-13 enzyme (40kDa) by western blot.

IHC: Human breast cancer Tissue, 1:20-1:200; Western Blot: MDA-MB-453s Cells, 1:200-1:2000; IF: MCF-7 Cells, 1:10-1:100

Type: Primary
Antigen: MMP13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR