Anti-PON1 Rabbit Polyclonal Antibody

Supplier: Proteintech
18155-1-AP-150UL
10092-588EA 402 USD
10092-588
Anti-PON1 Rabbit Polyclonal Antibody
Antibodies
PON1, also named as PON and K-45, belongs to the paraoxonase family. PON1 hydrolyzes the toxic metabolites of a variety of organophosphorus insecticides. It can capable of hydrolyzing a broad spectrum of organophosphate substrates and a number of aromatic carboxylic acid esters. PON1 may mediate an enzymatic protection of low density lipoproteins against oxidative modification and the consequent series of events leading to atheroma formation. For glycosylated, the MW of PON1 is migrated 42-45kd. 55kd is a dimmer of isoform CRA-b. The antibody has crss-reaction to PON2 and PON3.

Western Blot: Mouse Liver Tissue, 1:200-1:2000; IHC: Human Liver, 1:20-1:200

Type: Primary
Antigen: PON1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR