Anti-PODX Rabbit Polyclonal Antibody

Supplier: Proteintech
18150-1-AP-150UL
10092-534EA 537.7 USD
10092-534
Anti-PODX Rabbit Polyclonal Antibody
Antibodies
Podocalyxin, also known as podocalyxin-like protein 1 (PODXL or PCLP1), is a transmembrane glycoprotein belonging to the CD34 family of sialomucins. Podocalyxin was originally identified as the major sialoprotein on podocytes of the kidney glomerulus, but was later found to be expressed on vascular endothelial cells and early hematopoietic progenitors. It Involved in the regulation of both adhesion and cell morphology. In addition, podocalyxin is highly expressed in embryonic stem cells and aberrant expression of podocalyxin has been implicated in a wide range of cancers.

Western Blot: HEK-293 Cells, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200; FC: HeLa Cells, N/A

Type: Primary
Antigen: PODX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR