Anti-ABCD1 Rabbit Polyclonal Antibody

Supplier: Proteintech
18138-1-AP-150UL
10082-344EA 402 USD
10082-344
Anti-ABCD1 Rabbit Polyclonal Antibody
Antibodies
ABCD1 (also known as ALDP) is a member of the ATP-binding cassette (ABC) transporter superfamily which functions as transporter for a wide variety of substrates. It localizes to the peroxisomal membrane. The exact function is not clear so far. Various mutations of ABCD1 cause X-linked adrenoleukodystrophy (X-ALD), an inherited neurodegenerative disease affecting the nervous system white matter and adrenal cortex.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: ABCD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR