Anti-PARP15 Rabbit Polyclonal Antibody

Supplier: Proteintech
18126-1-AP-150UL
10091-806EA 402 USD
10091-806
Anti-PARP15 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A431 Cells, 1:200-1:2000

Type: Primary
Antigen: PARP15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR